Learn More
Agilent Technologies FP SEAHR 96 300U PVDF 1.0 50CS
Supplier:  Agilent Technologies 201023100
Encompass
Seahorse polypropylene 96-well filter microplate with 1um HYDROPHOBIC polyvinylidene fluoride membrane on a polypropylene support, 300ul/well, short drip, case of 50, recommended receiver plate 201276-100
Catalog No. 50-995-948