Learn More
Agilent Technologies FP SEAHR 96 300U PVDF .45 50CS
Catalog No. 50995930
Encompass
Seahorse polypropylene 96-well filter microplate with 0.45um polyvinylidene fluoride membrane, 300ul/well, short drip, case of 50, recommended receiver plate 201276-100
Catalog No. 50-995-930 Supplier Agilent Technologies Supplier No. 200995100
Add to Cart
Add to Cart
Seahorse polypropylene 96-well filter microplate with 0.45um polyvinylidene fluoride membrane, 300ul/well, short drip, case of 50, recommended receiver plate 201276-100