Learn More
Agilent Technologies FP SEAHR 96 400U PVDF .45 25CS
Catalog No. 50995905
Encompass
Seahorse polypropylene 96-well filter microplate with 0.45um polyvinylidene fluoride membrane, 400ul/well, long drip, case of 25, recommended receiver plate 201276-100
Catalog No. 50-995-905 Supplier Agilent Technologies Supplier No. 200943100
Add to Cart
Add to Cart
Seahorse polypropylene 96-well filter microplate with 0.45um polyvinylidene fluoride membrane, 400ul/well, long drip, case of 25, recommended receiver plate 201276-100