Learn More
Agilent Technologies FP SEAHR 96 400U PVDF .45 25CS
Supplier: Agilent Technologies 200971100
Seahorse polypropylene 96-well filter microplate with 0.45um polyvinylidene fluoride membrane, 400ul/well, short drip, case of 25, recommended receiver plate 201276-100