Learn More
Agilent Technologies Agilent filter microplate, 96-well, polypropylene, with 0.45 µm polyvinylidene fluoride membrane, 800 µL/well, long drip, 25/pk
Catalog No. 50995899
Encompass
Agilent filter microplate, 96-well, polypropylene, with 0.45 µm polyvinylidene fluoride membrane, 800 µL/well, long drip, 25/pk
Catalog No. 50-995-899 Supplier Agilent Technologies Supplier No. 200931100
Agilent filter microplate, 96-well, polypropylene, with 0.45 µm polyvinylidene fluoride membrane, 800 µL/well, long drip, 25/pk