Learn More
Agilent Technologies FP SEAHR 96 800U PVDF .45 25CS
Catalog No. 50995912
Encompass
Seahorse polypropylene 96-well filter microplate with 0.45um polyvinylidene fluoride membrane, 800ul/well, short drip, case of 25, recommended receiver plate 201276-100
Catalog No. 50-995-912 Supplier Agilent Technologies Supplier No. 200959100
Add to Cart
Add to Cart
Seahorse polypropylene 96-well filter microplate with 0.45um polyvinylidene fluoride membrane, 800ul/well, short drip, case of 25, recommended receiver plate 201276-100