Learn More
Agilent Technologies FP SEAHRS 384 140U GF 1.2 10CS
Supplier:  Agilent Technologies 201025100
Encompass
Seahorse polypropylene 384-well filter microplate with 1.2um glass fiber membrane, 140ul/well, long drip, case of 10, recommended receiver plate 201274-100
Catalog No. 50-995-949