Learn More
Agilent Technologies Agilent filter microplate, 384-well, polypropylene, with 0.7 µm glass fiber membrane, 140 µL/well, long drip, 10/pk
Supplier:  Agilent Technologies 201027100
Encompass
Agilent filter microplate, 384-well, polypropylene, with 0.7 µm glass fiber membrane, 140 µL/well, long drip, 10/pk
Catalog No. 50-995-950