Learn More
Agilent Technologies FP SEAHRSE 96 2ML GF .7 25/CS
Catalog No. 50995937
Encompass
Seahorse polypropylene 96-well filter microplate with 0.7um glass fiber membrane, 2ml/well, long drip, case of 25, recommended receiver plate 201240-100
Catalog No. 50-995-937 Supplier Agilent Technologies Supplier No. 201007100
Add to Cart
Add to Cart
Seahorse polypropylene 96-well filter microplate with 0.7um glass fiber membrane, 2ml/well, long drip, case of 25, recommended receiver plate 201240-100