Learn More
Agilent Technologies FP SEAHRSE 96 2ML PE 25 25/CS
Catalog No. 50995936
Encompass
Seahorse polypropylene 96-well filter microplate with 25um high molecular weight polyethylene membrane, 2ml/well, long drip, case of 25, recommended receiver plate 201240-100
Catalog No. 50-995-936 Supplier Agilent Technologies Supplier No. 201005100
Add to Cart
Add to Cart
Seahorse polypropylene 96-well filter microplate with 25um high molecular weight polyethylene membrane, 2ml/well, long drip, case of 25, recommended receiver plate 201240-100