Learn More
Agilent Technologies FP SEAHRSE 96 300U GF 1.0 50CS
Supplier:  Agilent Technologies 200985100
Encompass
Seahorse polypropylene 96-well filter microplate with 1.0um glass fiber membrane, 300ul/well, long drip, case of 50, recommended receiver plate 201276-100
Catalog No. 50-995-926