Learn More
Agilent Technologies FP SEAHRSE 96 300U GF 3.0 50CS
Catalog No. 50995944
Encompass
Seahorse polypropylene 96-well filter microplate with 3um glass fiber membrane, 300ul/well, long drip, case of 50, recommended receiver plate 201276-100
Catalog No. 50-995-944 Supplier Agilent Technologies Supplier No. 201021100
Add to Cart
Add to Cart
Seahorse polypropylene 96-well filter microplate with 3um glass fiber membrane, 300ul/well, long drip, case of 50, recommended receiver plate 201276-100