Learn More
Agilent Technologies FP SEAHRSE 96 300U GF .7 50CS
Catalog No. 50995933
Encompass
Seahorse polypropylene 96-well filter microplate with 0.7um glass fiber membrane, 300ul/well, short drip, case of 50, recommended receiver plate 201276-100
Catalog No. 50-995-933 Supplier Agilent Technologies Supplier No. 201001100
Add to Cart
Add to Cart
Seahorse polypropylene 96-well filter microplate with 0.7um glass fiber membrane, 300ul/well, short drip, case of 50, recommended receiver plate 201276-100