Learn More
Agilent Technologies FP SEAHRSE 96 300U PP .45 50CS
Supplier:  Agilent Technologies 200997100
Encompass
Seahorse polypropylene 96-well filter microplate with 0.45um polypropylene membrane, 300ul/well, short drip, case of 50, recommended receiver plate 201276-100
Catalog No. 50-995-931