Learn More
Agilent Technologies FP SEAHRSE 96 400U GF 1.2 25CS
Supplier:  Agilent Technologies 200941100
Encompass
Seahorse polypropylene 96-well filter microplate with 1.2um glass fiber membrane, 400ul/well, long drip, case of 25, recommended receiver plate 201276-100
Catalog No. 50-995-904