Learn More
Agilent Technologies FP SEAHRSE 96 400U PE 25 25/CS
Supplier:  Agilent Technologies 200939100
Encompass
Seahorse polypropylene 96-well filter microplate with 25um high molecular weight polyethylene membrane, 400ul/well, long drip, case of 25, recommended receiver plate 201276-100
Catalog No. 50-995-903