Learn More
Agilent Technologies FP SEAHRSE 96 400U PE 25 25CS
Catalog No. 50995916
Encompass
Seahorse polypropylene 96-well filter microplate with 25um high molecular weight polyethylene membrane, 400ul/well, short drip, case of 25, recommended receiver plate 201276-100
Catalog No. 50-995-916 Supplier Agilent Technologies Supplier No. 200967100
Add to Cart
Add to Cart
Seahorse polypropylene 96-well filter microplate with 25um high molecular weight polyethylene membrane, 400ul/well, short drip, case of 25, recommended receiver plate 201276-100