Learn More
Agilent Technologies FP SEAHRSE 96 400U PP .45 25CS
Supplier:  Agilent Technologies 200945100
Encompass
Seahorse polypropylene 96-well filter microplate with 0.45um polypropylene membrane, 400ul/well, long drip, case of 25, recommended receiver plate 201276-100
Catalog No. 50-995-906