Learn More
Agilent Technologies FP SEAHRSE 96 800U GF 1.0 25CS
Catalog No. 50995901
Encompass
Seahorse polypropylene 96-well filter microplate with 1.0um glass fiber membrane, 800ul/well, long drip, case of 25, recommended receiver plate 201276-100
Catalog No. 50-995-901 Supplier Agilent Technologies Supplier No. 200935100
Add to Cart
Add to Cart
Seahorse polypropylene 96-well filter microplate with 1.0um glass fiber membrane, 800ul/well, long drip, case of 25, recommended receiver plate 201276-100