Learn More
Agilent Technologies FP SEAHRSE 96 800U GF 1.2 25CS
Catalog No. 50995911
Encompass
Seahorse polypropylene 96-well filter microplate with 1.2um glass fiber membrane, 800ul/well, short drip, case of 25, recommended receiver plate 201276-100
Catalog No. 50-995-911 Supplier Agilent Technologies Supplier No. 200957100
Seahorse polypropylene 96-well filter microplate with 1.2um glass fiber membrane, 800ul/well, short drip, case of 25, recommended receiver plate 201276-100