Learn More
Agilent Technologies FP SEAHRSE 96 800U GF .7 25/CS
Catalog No. 50995902
Encompass
Seahorse polypropylene 96-well filter microplate with 0.7um glass fiber membrane, 800ul/well, long drip, case of 25, recommended receiver plate 201276-100
Catalog No. 50-995-902 Supplier Agilent Technologies Supplier No. 200937100
Add to Cart
Add to Cart
Seahorse polypropylene 96-well filter microplate with 0.7um glass fiber membrane, 800ul/well, long drip, case of 25, recommended receiver plate 201276-100