Learn More
Agilent Technologies Agilent filter microplate, 96-well, polypropylene, with 25 µm polyethylene frit, 800 µL/well, long drip, 25/pk
Catalog No. 50995909
Encompass
Agilent filter microplate, 96-well, polypropylene, with 25 µm polyethylene frit, 800 µL/well, long drip, 25/pk
Catalog No. 50-995-909 Supplier Agilent Technologies Supplier No. 200953100
Add to Cart
Add to Cart
Agilent filter microplate, 96-well, polypropylene, with 25 µm polyethylene frit, 800 µL/well, long drip, 25/pk