Learn More
Agilent Technologies Agilent filter microplate, 96-well, polypropylene, with 0.45 µm polypropylene membrane, 800 µL/well, long drip, 25/pk
Supplier:  Agilent Technologies 200933100
Encompass
Agilent filter microplate, 96-well, polypropylene, with 0.45 µm polypropylene membrane, 800 µL/well, long drip, 25/pk
Catalog No. 50-995-900