Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals FP96 BigFlex Metal Adapter

Catalog No. MP116010670 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Holds:
8 x 50 mL Tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
MP116010670 8 x 50 mL Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116010670 Supplier MP Biomedicals Supplier No. 116010670
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Securely holds up to 8 x 50 mL tubes. Resistant to a wide range of temperatures, suitable for various sample preparation conditions.

  • FP96 BigFlex Metal Adapter is a large-scale adapter for the FastPrep-96™ and 96™ Pro that holds 8 x 50 mL Lysing Matrix tubes (compatible with SafTest™ Food Inspection System).
  • A wide range of disposable 50 mL Lysing Matrix tubes ensure thorough homogenization of any sample type in seconds.
  • Made from lightweight anodized aluminum, it offers exceptional durability and consistency during homogenization.
  • Frozen samples can be directly loaded into the adapter for immediate processing, minimizing hands-on steps and reducing the risk of cellular degradation.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Adapter
Blade Material Stainless Steel
Color Blue
For Use With (Equipment) FastPrep-96, FastPrep-96 Pro
Holds 8 x 50 mL Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.