Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals FP96 TallFlex Metal Adapter

Catalog No. MP116010685 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Holds:
48 x 4.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
MP116010685 48 x 4.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116010685 Supplier MP Biomedicals Supplier No. 116010685
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Securely holds up to 48 x 4.5 mL tubes. Resistant to a wide range of temperatures, suitable for various sample preparation conditions.

  • Medium-scale FP96 TallFlex Metal Adapter for the FastPrep-96™ and FastPrep-96™ Pro that holds 48 x 4.5 mL Lysing Matrix Tubes.
  • The ergonomic design of the adapter ensures an easy loading of Lysing Matrix tubes that remain securely sealed during the homogenization.
  • It stands stable on the bench top and is commonly used as a tube rack.
  • Additionally, frozen samples in Lysing Matrix tubes loaded in the adapter and stored at -20 °C or -80 °C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.
  • Made from durable anodized aluminum for long-lasting performance.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Adapter
Blade Material Stainless Steel
Color Blue
For Use With (Equipment) FastPrep-96, FastPrep-96 Pro
Holds 48 x 4.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.