Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FPA 124, Tocris Bioscience™

Catalog No. 292610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-261-0 10 mg
29-265-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 29-261-0 Supplier R&D Systems Supplier No. 2926/10
This item is not returnable. View return policy

Akt/PKB inhibitor

Akt/PKB inhibitor (IC50 = 100 nM) that interacts with the PH and kinase domains. Inhibits cell proliferation in various cancer cell lines in vitro and decreases NF-κB activity and tumor load in vivo.

Chemical Identifiers

CAS 902779-59-3
Molecular Formula C11H9Cl2CuN3O2S
Molecular Weight (g/mol) 381.718
InChI Key HZRXQEVFXXQYFS-UHFFFAOYSA-L
Synonym Dichloro[(2Z)-2-[(4-oxo-4H-1-benzopyran-3-yl)methylene]hydrazinecarbothioamide copper complex
PubChem CID 102586612
IUPAC Name dichlorocopper;[(4-oxochromen-3-yl)methylideneamino]thiourea
SMILES C1=CC=C2C(=C1)C(=O)C(=CO2)C=NNC(=S)N.Cl[Cu]Cl

Specifications

Quantity 10 mg
Formula Weight 381.73
Percent Purity >98%
Chemical Name or Material FPA 124
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.