Learn More
Medchemexpress LLC HY-145319 5mg , FPFT-2216 CAS:2367619-87-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1453195MG
Encompass_Preferred
HY-145319 5mg FPFT-2216 CAS: 2367619-87-0 FPFT-2216, a “molecular glue” compound, degrades phosphodiesterase 6D (PDE6D), zinc finger transcription factors Ikaros (IKZF1), Aiolos (IKZF3), and casein kinase 1α (CK1α). FPFT-2216 can be used for the research of cancer and inflammatory disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1858