Learn More
Hoefer, Inc FPGD2 imaging System
SDP
Catalog No. NC3655196
Encompass_Preferred
"Flexible DNA, stain free, autorads analysisIntuitive touch screen systemCompact benchtop size with built in PCPowerfulA groundbreaking 5m pixel camera gives exquisite pixel resolution and unrivalled sensitivity in its class. FPGD2 uses an f/1.2 motor-driven zoom lens to enable perfect gel or blot-size imaging. The maximum viewing area is 20 x 24cm, which is very large for such a small, compact unit.FlexibleInternal lighting includes a UV transilluminator option for working with DNA gels, stain free and autorads analysis. The overhead LED white lighting is included as standard for easy sample positioning and focusing.VersatileThe FPGD2 has been designed for stain free applications. Its camera enables the sample to be exposed for extended periods. Safely excise your DNA bands from an agarose gel with the darkroom door open using the optional safety shield protecting the user when working with UV.
Catalog No. NC3655196 Supplier Hoefer, Inc Supplier No. FPGD2 SYSTEM
Add to Cart
Add to Cart
Request a Quote
"Flexible DNA, stain free, autorads analysisIntuitive touch screen systemCompact benchtop size with built in PCPowerfulA groundbreaking 5m pixel camera gives exquisite pixel resolution and unrivalled sensitivity in its class. FPGD2 uses an f/1.2 motor-driven zoom lens to enable perfect gel or blot-size imaging. The maximum viewing area is 20 x 24cm, which is very large for such a small, compact unit.FlexibleInternal lighting includes a UV transilluminator option for working with DNA gels, stain free and autorads analysis. The overhead LED white lighting is included as standard for easy sample positioning and focusing.VersatileThe FPGD2 has been designed for stain free applications. Its camera enables the sample to be exposed for extended periods. Safely excise your DNA bands from an agarose gel with the darkroom door open using the optional safety shield protecting the user when working with UV.