Learn More
Aviva Systems Biology Corp FPGT Antibody - C-terminal region (ARP62823_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62823_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FPGT. It was validated on Western Blot. . Alternate names: GFPPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rat, Yeast
Catalog No. 50-287-778