Learn More
TARGETMOL CHEMICALS INC FPH1 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T650625MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. FPH1 (BRD-6125) (BRD-6125) can promote the expansion of iPS-derived hepatocytes. purity: 99%
Catalog No. 50-257-9344