Learn More
Cayman Chemical FPL 55712sodIum salt 10mg
SDP
Catalog No. 502913935
Encompass_Preferred
An LT receptor antagonist; inhibits LTD4 binding to guinea pig lung homogenates (IC50 = 4 µM) and reduces LTD4-induced contraction of isolated guinea pig tracheal smooth muscle (pA2 = 1 µM); inhibits LTD4-, LTC4-, or LTE4-induced bronchoconstriction in guinea pigs (ID50s = 2, 6, and 1 mg/kg, respectively)
Catalog No. 50-291-3935 Supplier Cayman Chemical Supplier No. 3543310 MG
An LT receptor antagonist; inhibits LTD4 binding to guinea pig lung homogenates (IC50 = 4 µM) and reduces LTD4-induced contraction of isolated guinea pig tracheal smooth muscle (pA2 = 1 µM); inhibits LTD4-, LTC4-, or LTE4-induced bronchoconstriction in guinea pigs (ID50s = 2, 6, and 1 mg/kg, respectively)