Learn More
Medchemexpress LLC HY-105024 10mg Medchemexpress, FPL 62064 CAS:103141-09-9 Purity:>98%
SDP
Catalog No. 501968242
Encompass_Preferred
Medchemexpress, HY-105024 10mg FPL 62064 CAS:103141-09-9 FLP 62064 is a potent 5-lipoxygenase (5-LOX) and COX-2 dual inhibitor. FLP 62064 has potently anti-inflammatory activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8242 Supplier Medchemexpress LLC Supplier No. HY10502410MG
Medchemexpress, HY-105024 10mg FPL 62064 CAS:103141-09-9 FLP 62064 is a potent 5-lipoxygenase (5-LOX) and COX-2 dual inhibitor. FLP 62064 has potently anti-inflammatory activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.