Learn More
Medchemexpress LLC HY-105024 5mg Medchemexpress, FPL 62064 CAS:103141-09-9 Purity:>98%
SDP
Catalog No. 501968243
Encompass_Preferred
Medchemexpress, HY-105024 5mg FPL 62064 CAS:103141-09-9 FLP 62064 is a potent 5-lipoxygenase (5-LOX) and COX-2 dual inhibitor. FLP 62064 has potently anti-inflammatory activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8243 Supplier Medchemexpress LLC Supplier No. HY1050245MG
Medchemexpress, HY-105024 5mg FPL 62064 CAS:103141-09-9 FLP 62064 is a potent 5-lipoxygenase (5-LOX) and COX-2 dual inhibitor. FLP 62064 has potently anti-inflammatory activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.