Learn More
Sorbent Technologies Inc SorbaSep™ FPLC Desalting Columns, with Sorbadex S-25 Superfine for buffer exchange and/or desalting using a syringe, a pump, or a chromatography system. 5.0 ml, 5 columns
SDP
Catalog No. NC2963173
Encompass_Preferred
SorbaSep™ FPLC Desalting Columns - For desalting using FPLC and liquid chromatography systems (e.g. Cytiva AKTA™, Bio-Rad NGC™, and Knauer AZURA™). SorbaSep™ FPLC Columns are preloaded with SorbaDex™-25 Superfine, a beaded composite size-exclusion matrix. It exhibits high flow rates, excellent resolution, and chemical stability. The columns are used in a fractionation range for globular proteins from 1 and 5kD, aqueous compatible, pH range of 2-13.
Catalog No. NC2963173 Supplier Sorbent Technologies Inc Supplier No. 8030285
SorbaSep™ FPLC Desalting Columns - For desalting using FPLC and liquid chromatography systems (e.g. Cytiva AKTA™, Bio-Rad NGC™, and Knauer AZURA™). SorbaSep™ FPLC Columns are preloaded with SorbaDex™-25 Superfine, a beaded composite size-exclusion matrix. It exhibits high flow rates, excellent resolution, and chemical stability. The columns are used in a fractionation range for globular proteins from 1 and 5kD, aqueous compatible, pH range of 2-13.