Learn More
TARGETMOL CHEMICALS INC FPR A14 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T227895MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. FPR A14 is an agonist of formyl peptide receptor (FPR) and induces cell differentiation. purity: 99%
Catalog No. 50-258-0923