Learn More
Medchemexpress LLC HY-19574 5mg Medchemexpress, FPR Agonist 43 CAS:903895-98-7 Purity:>98%
SDP
Catalog No. 501932839
Encompass_Preferred
Medchemexpress, HY-19574 5mg FPR Agonist 43 CAS:903895-98-7 FPR Agonist 43 (compound 43) is a dual formyl peptide receptor 1 (FPR1) and formyl peptide receptor 2 (FPR2)/ALX agonist [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2839 Supplier Medchemexpress LLC Supplier No. HY195745MG
Medchemexpress, HY-19574 5mg FPR Agonist 43 CAS:903895-98-7 FPR Agonist 43 (compound 43) is a dual formyl peptide receptor 1 (FPR1) and formyl peptide receptor 2 (FPR2)/ALX agonist [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.