Learn More
Medchemexpress LLC HY-19370 5mg Medchemexpress, FPS-ZM1 CAS:945714-67-0 Purity:>98%
SDP
Catalog No. 502082128
Encompass_Preferred
Medchemexpress, HY-19370 5mg FPS-ZM1 CAS:945714-67-0 FPS-ZM1 is a high-affinity RAGE inhibitor with a Ki of 25 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2128 Supplier Medchemexpress LLC Supplier No. HY193705MG
Medchemexpress, HY-19370 5mg FPS-ZM1 CAS:945714-67-0 FPS-ZM1 is a high-affinity RAGE inhibitor with a Ki of 25 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.