Learn More
Medchemexpress LLC HY-100382 5mg Medchemexpress, FPTQ CAS:864863-72-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1003825MG
Encompass_Preferred
Medchemexpress, HY-100382 5mg FPTQ CAS:864863-72-9 FPTQ is potent mGluR1 antagonist with IC50 values of 6 nM and 1.4 nM for human and mouse mGluR1 respectively. FPTQ has anti-oxidant and anti-inflammatory effects in vitro and in vivo.
Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0584