Learn More
AdipoGen 3-(2-Furoyl)quinoline-2-carboxaldehyde, 126769-01-5, MFCD03427748, 10 mg
SDP
Catalog No. 501177468
Encompass_Preferred
MF: C15H9NO3, MW: 251.24, ≥95%, Appearance: White to off-white powder. 3-(2-Furoyl)quinoline-2-carboxaldehyde (FQCA) is a fluorogenic derivatizing reagent for liquid chromatography suitable for high-sensitivity analysis of amino acids by laser-induced fluorescence with an argon-ion laser. The reagent forms highly fluorescent isoindoles upon reaction with primary amines.
Catalog No. 50-117-7468 Supplier AdipoGen Supplier No. CDXF0001M010
MF: C15H9NO3, MW: 251.24, ≥95%, Appearance: White to off-white powder. 3-(2-Furoyl)quinoline-2-carboxaldehyde (FQCA) is a fluorogenic derivatizing reagent for liquid chromatography suitable for high-sensitivity analysis of amino acids by laser-induced fluorescence with an argon-ion laser. The reagent forms highly fluorescent isoindoles upon reaction with primary amines.