Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FR 122047 hydrochloride, Tocris Bioscience™

Catalog No. 150750
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-075-0 50 mg
15-071-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 15-075-0 Supplier R&D Systems Supplier No. 1507/50
This item is not returnable. View return policy

Cyclooxygenase (COX-1) inhibitor

Selective cyclooxygenase-1 (COX-1) inhibitor (IC50 values are 0.028 and 65 μM for COX-1 and COX-2 respectively). Antiplatelet, analgesic and anti-inflammatory following oral administration in vivo.

Chemical Identifiers

CAS 130717-51-0
Molecular Formula C23H26ClN3O3S
Molecular Weight (g/mol) 459.989
InChI Key YWMAVHIKOAOSFM-UHFFFAOYSA-N
Synonym fr 122047 hydrochloride, cox-1 inhibitor, fr122047, 1-4,5-bis 4-methoxyphenyl-2-thiazolyl carbonyl-4-methylpiperazine hydrochloride, 4,5-bis 4-methoxyphenyl-1,3-thiazol-2-yl-4-methylpiperazin-1-yl methanone hydrochloride, 1-4,5-bis 4-methoxyphenyl-2-thiazoyl carbonyl-4-methylpiperazine, hydrochloride, 4,5-bis 4-methoxyphenyl-2-1-methylpiperazin-4-yl carbonyl thiazole, hydrochloride, 1-4,5-bis 4-methoxyphenyl-1,3-thiazole-2-carbonyl-4-methylpiperazine hydrochloride, 1-4,5-bis 4-methoxyphenyl-2-thiazoyl carbonyl-4-methylpiperazine, d09cmj, piperazine, 1-4,5-bis 4-methoxyphenyl-2-thiazolyl carbonyl-4-methyl-, monohydrochloride
PubChem CID 196840
IUPAC Name [4,5-bis(4-methoxyphenyl)-1,3-thiazol-2-yl]-(4-methylpiperazin-1-yl)methanone;hydrochloride
SMILES CN1CCN(CC1)C(=O)C2=NC(=C(S2)C3=CC=C(C=C3)OC)C4=CC=C(C=C4)OC.Cl

Specifications

Quantity 50 mg
Formula Weight 459.99
Percent Purity >98%
Chemical Name or Material FR 122047 hydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.