Learn More
Medchemexpress LLC HY-U00146 1mg Medchemexpress, FR-188582 CAS:189699-82-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYU001461MG
Encompass_Preferred
Medchemexpress, HY-U00146 1mg FR-188582 CAS:189699-82-9 FR-188582 is a highly selective inhibitor of cyclooxygenase ( COX )-2, with an IC 50 value of 17 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4427