Learn More
Description
- Soft and breathable FR fabric
- Designed for comfort and flexibility
- Elastic fit for comfort
- FR fabric will not melt, drip, or ignite
- Full coverage
Specifications
Specifications
| Size | Universal |
| Material | Nomex |
| Color | Gray |
| Disposable | Reusable |
| Certifications/Compliance | NFPA 70E, ASTM F1506, CSA Z462, OSHA 1910.269 |
| For Use With (Application) | Food Processing, Manufacturing |
| Quantity | 1 Ea. |
| Product Type | Flame-resistant Beard Cover |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
