Learn More
Abcam Assay Kits, Enzyme substrates, Enzyme substrates for Peptidases, Enzyme substrates for Proteases, Magic RedTM-(FR)2 Cathepsin B Assay Kit - 25tests, Magic Red substrate-based assays are designed to detect Cathepsin protease activity within whole living cells, 1 kit
Catalog No. 50996171
Encompass_Preferred
Assay Kits, Enzyme substrates, Enzyme substrates for Peptidases, Enzyme substrates for Proteases, Magic RedTM-(FR)2 Cathepsin B Assay Kit - 25tests, Magic Red substrate-based assays are designed to detect Cathepsin protease activity within whole living cells, 1 kit
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-171 Supplier Abcam Supplier No. AB27077425TEST
Assay Kits, Enzyme substrates, Enzyme substrates for Peptidases, Enzyme substrates for Proteases, Magic RedTM-(FR)2 Cathepsin B Assay Kit - 25tests, Magic Red substrate-based assays are designed to detect Cathepsin protease activity within whole living cells, 1 kit
The product is subject to the following: Abcam Restricted Use Statement