Learn More
International Enviroguard™ FR and Chemical Resistant Coverall with Attached Hood and Elastic Wrists
Catalog No. 17184014 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-184-014 Medium
17-184-015 Large
17-184-013 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-184-014 Supplier International Enviroguard™ Supplier No. 9115TM
Add to Cart
Add to Cart
PyroGuard CRFR flame-resistant coveralls are designed to wear over your primary FR garments, helping to extend the life of your primary FR clothing.
  • Coveralls also offer protection from a broad range of chemicals and provide flash fire protection that meets NFPA 2113
  • Advanced, self-extinguishing fabric does not melt or drip when ignited to prevent flames from spreading
  • Attached respirator-fit hood combined with secure, elastic wrists accommodate a respirator to prevent toxic inhalation and keep sleeves in place to avoid skin exposure
  • PyroGuard CRFR flame-resistant suits stop flames and chemicals in their tracks offering the comfort and safety you need to work in tough environments
  • Proprietary bilaminate fabric protects against flames and chemicals
  • Elastic back stretches with user for more comfort and better tear resistance
  • Storm flap over zipper adds an extra layer of protection
  • Taped seams offer added protection over serged seam coveralls
  • Possible applications include petrochemical environments, manufacturing, industrial, oil/gas, and more
Specifications
Product Type Flame-resistant Coveralls
Size Medium
Gender Unisex
Material CFR Bilaminate Fabric
Color Gray
Disposable Single-use
Waist Style Elastic at Back
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached, Elastic Face
Closure Type 2-way Front Zipper with Storm Flap
Seam Type Serged, Taped
Sterility Non-sterile
Resistance Acid, Anti-static, Caustics, Chemicals, Flame
Latex Free Yes
No. of Pockets None