Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ FR Chemical Resistant Lab Coat , Royal Blue
SDP

Catalog No. 17010895 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-895 Large
17-010-893 Small
17-010-894 Medium
17-010-896 X-Large
17-010-897 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-010-895 Supplier National Safety Apparel™ Supplier No. LABCRRBLG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lab coat features proprietary CR/FR fabric to provide multi-hazard protection without feeling weighted. Breathable chemical resistance meets superior flame resistance for protection and does not sacrifice comfort.

  • FR Nomex™ knit wrists keep sleeves in place
  • Durable, high-temp plastic snaps for easy donning and doffing
  • Bottom side vents for increased mobility
  • Side pass-through allows access to pockets underneath the coat
  • Durable accessory loop for safety glasses, pens, badges, etc.
  • Chest pocket with covered flap and pen/pencil slot
  • Lower pockets with covered flaps to secure contents
  • Highly breathable compared to traditional chemical protective options
  • Easy care and laundering
  • 42 in. length

Specifications

Product Type Flame-resistant Lab Coat
Size Large
Gender Unisex
Material Aramid Fiber
Color Royal Blue
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Resistance Fluids, Flame
Style Standard Length
No. of Pockets 3
Certifications/Compliance NFPA 70E, NFPA 2112, ASTM F1506, CSA Z462, OSHA 1910.269
For Use With (Application) Manufacturing, Food Processing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.