Learn More
National Safety Apparel™ DRIFIRE™ FR Control™ Long Underwear
Shop All National Safety Apparel Products

Description
- Proactively regulates skin temperature (± 3 degrees)
- Reduces perspiration
- Maintains a comfortable work environment
- Keeps the wearer cooler on hot days, warmer on cool days
- Heat stress reduction
- Odor control technology
- Lightweight, breathable fabric
- Unsurpassed moisture wicking and drying rate
- FR fabric will not melt, drip, or ignite
- Inherently flame resistant
- FR protection will not wash/wear out
- 4-way stretch fabric for mobility and recovery
- Made in the USA
Specifications
Specifications
| Size | 3X-Large |
| Gender | Unisex |
| Product Type | Multirisk Pants |
| Material | Modacrylic |
| Color | Desert Sand |
| Seam Type | Stitched |
| No. of Pockets | None |
| Disposable | Reusable |
| Hazard Risk Category | CAT 1 |
| Arc Flash Rating | 4 cal/cm² |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.