Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ DRIFIRE™ FR Control ™ Long Sleeve T-Shirt
SDP

Catalog No. 17355134 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-355-134 Small
17-355-135 Medium
17-355-136 Large
17-355-137 X-Large
17-355-138 2X-Large
17-355-139 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-355-134 Supplier National Safety Apparel™ Supplier No. C52JKSRLSSM
Add to Cart
Add to Cart
This item is not returnable. View return policy

Long Sleeve T-shirt is designed to proactively regulate skin temperature.

  • Proactively regulates skin temperature (± 3 degrees)
  • Reduces perspiration by 34%
  • 4-way stretch fabric for mobility and recovery
  • Heat stress reduction
  • Odor control
  • Maintains comfortable temperature in hot and cold conditions
  • Superior moisture wicking and dry rate
  • Inherently flame resistant
  • FR protection will not wash/wear out
  • Lightweight, breathable fabric
  • Made in the USA

Specifications

Size Small
Gender Unisex
Material Modacrylic Blend
Color Desert Sand
Neck Style Crew
Sleeve Style Long Sleeve
Closure Type Pull On
Disposable Reusable
Product Type Shirt
Hazard Risk Category CAT 1
Arc Flash Rating 4 cal/cm²
Certifications/Compliance NFPA 70E, ASTM F1506, CSA Z462, OSHA 1910.269
Style Base Layer Shirt
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.