Learn More
Lakeland Industries FR Coverall
Catalog No. 502840777
Encompass_Preferred
Lakeland CAT 2 Inherently FR 9.0 oz. Cotton coveralls are built tough and comfortable to protect the wearers from flash fires. C081 coveralls are configured with a quick release zipper, heavy duty snaps, pockets, and an elasticized waistband. Available in Navy and Khaki with reflective trim options, sizes SM-6X, this coverall is ideal when arc flash and flash fire hazards are present.
Catalog No. 50-284-0777 Supplier Lakeland Industries Supplier No. C08113LG30
Lakeland CAT 2 Inherently FR 9.0 oz. Cotton coveralls are built tough and comfortable to protect the wearers from flash fires. C081 coveralls are configured with a quick release zipper, heavy duty snaps, pockets, and an elasticized waistband. Available in Navy and Khaki with reflective trim options, sizes SM-6X, this coverall is ideal when arc flash and flash fire hazards are present.