Learn More
National Safety Apparel™ FR Fleece 3-in-1 Hood
SDP
Catalog No. 17001744 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Color: Navy
Catalog No.
17-001-744 Color: Navy
1 options
Catalog No. 17-001-744 Supplier National Safety Apparel™ Supplier No. H74FL28
Designed for cold weather conditions, the FR Fleece 3-in-1 Hood combines a fleece hood with an adjustable inner facial liner and neck gaiter
  • Arc Rating: 24 cal/cm²
  • CAT 2
  • Fabric/Material: 8.5 oz. DuPont™ Nomex™ IIIA Fleece (Hood) / 5.5 oz. FR Control 2.0 (Neck gaiter/face cover)
  • Adjustable drawstring hood
  • Combines a fleece hood with an adjustable inner facial liner/neck gaiter
  • Several unique ways to wear
  • Warmth without the weight
  • Breathable and quick drying FR Fabric
  • Wicks moisture away from the skin for comfort and warmth (inner liner)
  • Designed for cold weather conditions
  • Made in the USA
Specifications
Size Medium
Material DuPont™ Nomex™, Kevlar™, Anti-Stat, Modacrylic, Viscose, Elastane
Color Navy
Seam Type Stitched
Arc Flash Rating 24 cal/cm²
Certifications/Compliance CAT 2, NFPA 70 E, ASTM F1506, CSA Z462, OSHA 1910.269