Learn More
National Safety Apparel™ FR Fleece Neck Gaiter
SDP
Catalog No. 17001697 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Color: Navy
Catalog No.
17-001-697 Color: Navy
1 options
Catalog No. 17-001-697 Supplier National Safety Apparel™ Supplier No. H01FL159
DuPont™ Nomex™ brand fiber provides lightweight, durable, heat and flame resistant solutions. Made of 93% Nomex, 5% Kevlar, and 2% anti-static materials, Nomex™ IIIA is engineered to absorb moisture and dry quickly
  • Arc Rating: 24 cal/cm²
  • CAT 2
  • Fabric/Material: 8.5 oz. DuPont™ Nomex™ IIIA Fleece
  • Designed for cold weather conditions
  • Warmth without the weight
  • FR fabric will not melt, drip or ignite
  • Made in the USA
Specifications
Size Medium
Material DuPont™ Nomex™ IIIA Fleece
Color Navy
Seam Type Stitched
Arc Flash Rating 24 cal/cm²
Certifications/Compliance CAT 2, ASTM F1506, OSHA 1910.269, NFPA 70 E, NFPA 2112, CSA Z462